Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-heptynoate, 98+%

$38.95 - $133.92

Chemical Identifiers

Chemical Name or Material Ethyl 2-heptynoate
CAS 16930-95-3
Molecular Formula C9H14O2
Linear Formula CH3(CH2)3C≡CCOOCH2CH3
Formula Weight 154.21
MDL Number MFCD00015275
Beilstein 1756846
Synonym 2-Heptynoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0274903

  SDS
alfa aesar™
L0274903
1g Each for $38.95

AAL0274906

  SDS
alfa aesar™
L0274906
5g Each for $133.92
Description & Specifications

Specifications

Formula Weight 154.21
Chemical Name or Material Ethyl 2-heptynoate
Assay ≥98%
Molecular Formula C9H14O2
Linear Formula CH3(CH2)3C≡CCOOCH2CH3
MDL Number MFCD00015275
CAS 16930-95-3
Beilstein 1756846
Synonym 2-Heptynoic acid ethyl ester
Health Hazard 1 Warning
Odor Recognizable
Boiling Point 193° to 194°C
Quantity 1g
Recommended Storage 2° to 8°C
Safety and Handling

Warning

2° to 8°C