Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl (2-formylphenoxy)acetate, 99%

$86.66 - $292.46

Chemical Identifiers

Chemical Name or Material Ethyl (2-formylphenoxy)acetate
CAS 41873-61-4
Molecular Formula C11H12O4
Formula Weight 208.22
MDL Number MFCD00052131
Synonym (2-Formylphenoxy)acetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5028603

 
alfa aesar™
H5028603
1g Each for $86.66

AAH5028606

 
alfa aesar™
H5028606
5g Each for $292.46
Description & Specifications

Specifications

Formula Weight 208.22
Chemical Name or Material Ethyl (2-formylphenoxy)acetate
Assay 99%
Molecular Formula C11H12O4
MDL Number MFCD00052131
CAS 41873-61-4
Synonym (2-Formylphenoxy)acetic acid ethyl ester
Quantity 1g