missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 2-Fluoropropionate 97.0+%, TCI America™
$76.32 - $210.70
Chemical Identifiers
CAS | 349-43-9 |
---|---|
Molecular Formula | C5H9FO2 |
Molecular Weight (g/mol) | 120.12 |
MDL Number | MFCD01320801 |
InChI Key | ODMITNOQNBVSQG-UHFFFAOYNA-N |
Synonym | 2-Fluoropropionic Acid Ethyl Ester |
PubChem CID | 265947 |
IUPAC Name | ethyl 2-fluoropropanoate |
SMILES | CCOC(=O)C(C)F |
Chemical Identifiers
349-43-9 | |
120.12 | |
ODMITNOQNBVSQG-UHFFFAOYNA-N | |
265947 | |
CCOC(=O)C(C)F |
C5H9FO2 | |
MFCD01320801 | |
2-Fluoropropionic Acid Ethyl Ester | |
ethyl 2-fluoropropanoate |
Specifications
349-43-9 | |
121°C | |
MFCD01320801 | |
3272 | |
ODMITNOQNBVSQG-UHFFFAOYNA-N | |
ethyl 2-fluoropropanoate | |
265947 | |
≥97.0% (GC) | |
Ethyl 2-Fluoropropionate |
Yellow | |
C5H9FO2 | |
5 g | |
2-Fluoropropionic Acid Ethyl Ester | |
CCOC(=O)C(C)F | |
120.12 | |
120.12 | |
Liquid |
Safety and Handling
TSCA : No