Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-fluorophenylacetate, 99%

$46.26 - $149.92

Chemical Identifiers

Chemical Name or Material Ethyl 2-fluorophenylacetate
CAS 584-74-7
Molecular Formula C10H11FO2
Formula Weight 182.2
MDL Number MFCD04039343
Synonym 2-Fluorophenylacetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1999906

  SDS
alfa aesar™
L1999906
5g Each for $46.26

AAL1999914

  SDS
alfa aesar™
L1999914
25g Each for $149.92
Description & Specifications

Specifications

Formula Weight 182.2
Chemical Name or Material Ethyl 2-fluorophenylacetate
Molecular Formula C10H11FO2
MDL Number MFCD04039343
CAS 584-74-7
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 99%
Synonym 2-Fluorophenylacetic acid ethyl ester
Boiling Point 235°C
Health Hazard 1 Warning
Quantity 5g
Flash Point 85°C (185°F)
Safety and Handling

Warning

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.