missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl (2-Fluorobenzoyl)acetate, TCI America™

Manufacturer:  TCI America E10975G

 View more versions of this product

Catalog No. E10975G



Specifications

Specifications

Ethyl (2-Fluorobenzoyl)acetate
Yellow
1479-24-9
MFCD00000321
YMUNUVJSNWUWDA-UHFFFAOYSA-N
ethyl 3-(2-fluorophenyl)-3-oxopropanoate
296342
Clear Liquid at 20°C
108°C
5g
C11H11FO3
ethyl 3-2-fluorophenyl-3-oxopropanoate, ethyl 2-fluorobenzoyl acetate, ethyl 3-2-fluorophenyl-3-oxo-propionate, 3-2-fluoro-phenyl-3-oxo-propionic acid ethyl ester, ethyl 3-2-fluorophenyl-3-oxopropionate, 3-2-fluorophenyl-3-oxopropionic acid ethyl ester, benzenepropanoic acid, 2-fluoro-beta-oxo-, ethyl ester, acmc-1c1es, ethyl 2-fluorobenzoylacetate, ethyl-2-fluorobenzoylacetate
CCOC(=O)CC(=O)C1=CC=CC=C1F
210.204
210.20
≥98.0% (GC)

Chemical Identifiers

1479-24-9
210.204
YMUNUVJSNWUWDA-UHFFFAOYSA-N
296342
CCOC(=O)CC(=O)C1=CC=CC=C1F
C11H11FO3
MFCD00000321
ethyl 3-2-fluorophenyl-3-oxopropanoate, ethyl 2-fluorobenzoyl acetate, ethyl 3-2-fluorophenyl-3-oxo-propionate, 3-2-fluoro-phenyl-3-oxo-propionic acid ethyl ester, ethyl 3-2-fluorophenyl-3-oxopropionate, 3-2-fluorophenyl-3-oxopropionic acid ethyl ester, benzenepropanoic acid, 2-fluoro-beta-oxo-, ethyl ester, acmc-1c1es, ethyl 2-fluorobenzoylacetate, ethyl-2-fluorobenzoylacetate
ethyl 3-(2-fluorophenyl)-3-oxopropanoate
Safety and Handling

Safety and Handling

TSCA : No

Documents