Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl (2-fluorobenzoyl)acetate, 97%

Chemical Identifiers

Chemical Name or Material Ethyl (2-fluorobenzoyl)acetate
CAS 1479-24-9
Formula Weight 210.2
MDL Number MFCD00000321
Synonym 2-Fluorobenzoylacetic acid ethyl ester, 97%
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6120403

 
alfa aesar™
H6120403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6120406

 
alfa aesar™
H6120406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 210.2
Chemical Name or Material Ethyl (2-fluorobenzoyl)acetate
Density 1.164g/mL
Physical Form Liquid
Assay 97%
Refractive Index 1.5160
MDL Number MFCD00000321
CAS 1479-24-9
Synonym 2-Fluorobenzoylacetic acid ethyl ester, 97%
Quantity 1g