Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-fluorobenzoate, 98+%

$84.97 - $278.61

Chemical Identifiers

Chemical Name or Material Ethyl 2-fluorobenzoate
CAS 443-26-5
Molecular Formula C9H9FO2
Formula Weight 168.17
MDL Number MFCD00039214
Beilstein 2084296
Synonym 2-Fluorobenzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1656414

 
alfa aesar™
A1656414
25g Each for $84.97

AAA1656422

 
alfa aesar™
A1656422
100g Each for $278.61
Description & Specifications

Specifications

Formula Weight 168.17
Chemical Name or Material Ethyl 2-fluorobenzoate
TSCA TSCA
Assay ≥98%
Molecular Formula C9H9FO2
MDL Number MFCD00039214
CAS 443-26-5
Beilstein 2084296
Synonym 2-Fluorobenzoic acid ethyl ester
Boiling Point 217° to 220°C
Quantity 25g