Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-ethylbutyrate, 99%

Chemical Identifiers

Chemical Name or Material Ethyl 2-ethylbutyrate
CAS 2983-38-2
Molecular Formula (CH3CH2)2CHCO2CH2CH3
Formula Weight 144.21
MDL Number MFCD00059396
UN Number UN3272
Synonym 2-Ethylbutyric acid ethyl ester; Ethyl diethylacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2578806

  SDS
alfa aesar™
H2578806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2578814

  SDS
alfa aesar™
H2578814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 144.21
Molecular Formula (CH3CH2)2CHCO2CH2CH3
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
MDL Number MFCD00059396
Quantity 5g
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Melting Point 149° to 152°C
Assay 99%
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 39°C (102°F)
UN Number UN3272
Synonym 2-Ethylbutyric acid ethyl ester; Ethyl diethylacetate
CAS 2983-38-2
Chemical Name or Material Ethyl 2-ethylbutyrate
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.