missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 2-(ethoxycarbonylmethoxy)benzoate, 97%, Thermo Scientific™
$295.30 - $295.30
Chemical Identifiers
CAS | 56424-77-2 |
---|---|
Molecular Formula | C13H16O5 |
Molecular Weight (g/mol) | 252.266 |
MDL Number | MFCD00151820 |
InChI Key | NIUVYKIVCCMXLR-UHFFFAOYSA-N |
Synonym | ethyl 2-2-ethoxy-2-oxoethoxy benzoate, ethyl 2-ethoxycarbonylmethoxy benzoate, ethyl o-carboethoxymethylsalicylate, ethyl 2-2-ethoxy-2-oxo-ethoxy benzoate, ethyl 2-2-ethoxycarbonyl phenoxy acetate, 2-2-ethoxy-2-oxoethoxy benzoic acid ethyl ester, ethyl 2-2-ethoxy-2-oxidanylidene-ethoxy benzoate, benzoic acid,2-2-ethoxy-2-oxoethoxy-, ethyl ester |
PubChem CID | 347810 |
IUPAC Name | ethyl 2-(2-ethoxy-2-oxoethoxy)benzoate |
SMILES | CCOC(=O)COC1=CC=CC=C1C(=O)OCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1343018
|
Thermo Scientific Chemicals
L1343018 |
50 g |
Each for $295.30
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
56424-77-2 | |
252.266 | |
NIUVYKIVCCMXLR-UHFFFAOYSA-N | |
347810 | |
CCOC(=O)COC1=CC=CC=C1C(=O)OCC |
C13H16O5 | |
MFCD00151820 | |
ethyl 2-2-ethoxy-2-oxoethoxy benzoate, ethyl 2-ethoxycarbonylmethoxy benzoate, ethyl o-carboethoxymethylsalicylate, ethyl 2-2-ethoxy-2-oxo-ethoxy benzoate, ethyl 2-2-ethoxycarbonyl phenoxy acetate, 2-2-ethoxy-2-oxoethoxy benzoic acid ethyl ester, ethyl 2-2-ethoxy-2-oxidanylidene-ethoxy benzoate, benzoic acid,2-2-ethoxy-2-oxoethoxy-, ethyl ester | |
ethyl 2-(2-ethoxy-2-oxoethoxy)benzoate |
Specifications
56424-77-2 | |
C13H16O5 | |
MFCD00151820 | |
2659411 | |
NIUVYKIVCCMXLR-UHFFFAOYSA-N | |
ethyl 2-(2-ethoxy-2-oxoethoxy)benzoate | |
347810 | |
97% |
180°C (4 mmHg) | |
1.514 | |
50 g | |
ethyl 2-2-ethoxy-2-oxoethoxy benzoate, ethyl 2-ethoxycarbonylmethoxy benzoate, ethyl o-carboethoxymethylsalicylate, ethyl 2-2-ethoxy-2-oxo-ethoxy benzoate, ethyl 2-2-ethoxycarbonyl phenoxy acetate, 2-2-ethoxy-2-oxoethoxy benzoic acid ethyl ester, ethyl 2-2-ethoxy-2-oxidanylidene-ethoxy benzoate, benzoic acid,2-2-ethoxy-2-oxoethoxy-, ethyl ester | |
CCOC(=O)COC1=CC=CC=C1C(=O)OCC | |
252.266 | |
252.27 | |
Ethyl 2-(ethoxycarbonylmethoxy)benzoate |
Safety and Handling
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H302-H315-H319
TSCA : No
Recommended Storage : Ambient temperatures