missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-oxocyclohexanecarboxylate, 95%, ACROS Organics™

Chemical Identifiers

CAS 1655-07-8
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.21
MDL Number MFCD00001631
InChI Key FGSGHBPKHFDJOP-UHFFFAOYSA-N
Synonym 2-carbethoxycyclohexanone, 2-cyclohexanonecarboxylic acid ethyl ester, 2-ethoxycarbonyl cyclohexanone, 2-oxocyclohexanecarboxylic acid ethyl ester, cyclohexanecarboxylic acid, 2-oxo-, ethyl ester, ethyl 2-cyclohexanonecarboxylate, ethyl 2-oxo-1-cyclohexanecarboxylate, ethyl 2-oxocyclohexanecarboxylate, ethyl 2-oxocylohexanecarboxylate, ethyl cyclohexanone-2-carboxylate
PubChem CID 95543
IUPAC Name ethyl 2-oxocyclohexane-1-carboxylate
SMILES CCOC(=O)C1CCCCC1=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC118330050
SDS View Documents
Fisher Scientific Acros
118330050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1655-07-8
170.21
FGSGHBPKHFDJOP-UHFFFAOYSA-N
95543
CCOC(=O)C1CCCCC1=O
C9H14O3
MFCD00001631
2-carbethoxycyclohexanone, 2-cyclohexanonecarboxylic acid ethyl ester, 2-ethoxycarbonyl cyclohexanone, 2-oxocyclohexanecarboxylic acid ethyl ester, cyclohexanecarboxylic acid, 2-oxo-, ethyl ester, ethyl 2-cyclohexanonecarboxylate, ethyl 2-oxo-1-cyclohexanecarboxylate, ethyl 2-oxocyclohexanecarboxylate, ethyl 2-oxocylohexanecarboxylate, ethyl cyclohexanone-2-carboxylate
ethyl 2-oxocyclohexane-1-carboxylate

Specifications

1.0600g/mL
Glass bottle
1.06
5g
94% min. (GC)
(O=)C6H9CO2C2H5
10, IV, 2606
FGSGHBPKHFDJOP-UHFFFAOYSA-N
ethyl 2-oxocyclohexane-1-carboxylate
95543
95%
85°C
1.4775 to 1.4795
106.0°C (11.0 mmHg)
1655-07-8
C9H14O3
MFCD00001631
2-carbethoxycyclohexanone, 2-cyclohexanonecarboxylic acid ethyl ester, 2-ethoxycarbonyl cyclohexanone, 2-oxocyclohexanecarboxylic acid ethyl ester, cyclohexanecarboxylic acid, 2-oxo-, ethyl ester, ethyl 2-cyclohexanonecarboxylate, ethyl 2-oxo-1-cyclohexanecarboxylate, ethyl 2-oxocyclohexanecarboxylate, ethyl 2-oxocylohexanecarboxylate, ethyl cyclohexanone-2-carboxylate
CCOC(=O)C1CCCCC1=O
170.21
170.21
Ethyl 2-oxocyclohexanecarboxylate, 98%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 216-731-5

SDS