missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-cyanobutyrate, 97%

$89.45 - $285.30

Chemical Identifiers

CAS 1619-58-5
Molecular Formula C7H11NO2
Molecular Weight (g/mol) 141.17
MDL Number MFCD02258598
InChI Key FCISHUHNFYJJDU-UHFFFAOYSA-N
Synonym ethyl2-cyanobutanoate, butanoic acid, 2-cyano-, ethyl ester, ethyl 2-cyanobutyrate, ksc531a2r, 2-cyanobutyric acid ethyl ester, 2-cyanobutanoic acid ethyl ester, 2-cyano-butyric acid ethyl ester, butanoic acid,2-cyano-, ethyl ester, 2-cyanobutyric acid, ethyl ester
PubChem CID 219611
IUPAC Name ethyl 2-cyanobutanoate
SMILES CCC(C#N)C(=O)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH65626MD View Documents Alfa Aesar™
H65626MD
250mg Each for $89.45
 
AAH6562603 View Documents Alfa Aesar™
H6562603
1g Each for $285.30
 
Specifications

Chemical Identifiers

1619-58-5
141.17
FCISHUHNFYJJDU-UHFFFAOYSA-N
219611
CCC(C#N)C(=O)OCC
C7H11NO2
MFCD02258598
ethyl2-cyanobutanoate, butanoic acid, 2-cyano-, ethyl ester, ethyl 2-cyanobutyrate, ksc531a2r, 2-cyanobutyric acid ethyl ester, 2-cyanobutanoic acid ethyl ester, 2-cyano-butyric acid ethyl ester, butanoic acid,2-cyano-, ethyl ester, 2-cyanobutyric acid, ethyl ester
ethyl 2-cyanobutanoate

Specifications

0.985g/mL
Colorless
1619-58-5
MFCD02258598
385926
FCISHUHNFYJJDU-UHFFFAOYSA-N
ethyl 2-cyanobutanoate
219611
Liquid
Ethyl 2-cyanobutyrate
1.4170
250mg
C7H11NO2
UN3276
ethyl2-cyanobutanoate, butanoic acid, 2-cyano-, ethyl ester, ethyl 2-cyanobutyrate, ksc531a2r, 2-cyanobutyric acid ethyl ester, 2-cyanobutanoic acid ethyl ester, 2-cyano-butyric acid ethyl ester, butanoic acid,2-cyano-, ethyl ester, 2-cyanobutyric acid, ethyl ester
CCC(C#N)C(=O)OCC
141.17
141.17
97%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.