Learn More
Ethyl 2-cyanobutyrate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H65626MD
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ethyl 2-cyanobutyrate | |
Colorless | |
83°C (181°F) | |
1.417 | |
250 mg | |
385926 | |
FCISHUHNFYJJDU-UHFFFAOYSA-N | |
ethyl 2-cyanobutanoate | |
219611 | |
97% |
1619-58-5 | |
0.985 g/mL | |
C7H11NO2 | |
MFCD02258598 | |
UN3276 | |
ethyl2-cyanobutanoate, butanoic acid, 2-cyano-, ethyl ester, ethyl 2-cyanobutyrate, ksc531a2r, 2-cyanobutyric acid ethyl ester, 2-cyanobutanoic acid ethyl ester, 2-cyano-butyric acid ethyl ester, butanoic acid,2-cyano-, ethyl ester, 2-cyanobutyric acid, ethyl ester | |
CCC(C#N)C(=O)OCC | |
141.17 | |
141.17 | |
Liquid |
Chemical Identifiers
1619-58-5 | |
141.17 | |
FCISHUHNFYJJDU-UHFFFAOYSA-N | |
219611 | |
CCC(C#N)C(=O)OCC |
C7H11NO2 | |
MFCD02258598 | |
ethyl2-cyanobutanoate, butanoic acid, 2-cyano-, ethyl ester, ethyl 2-cyanobutyrate, ksc531a2r, 2-cyanobutyric acid ethyl ester, 2-cyanobutanoic acid ethyl ester, 2-cyano-butyric acid ethyl ester, butanoic acid,2-cyano-, ethyl ester, 2-cyanobutyric acid, ethyl ester | |
ethyl 2-cyanobutanoate |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1
TSCA : No
Recommended Storage : Ambient temperatures