Please login to your online account to display your discounted pricing

ethyl 2-cyanoacetate, Maybridge

Chemical Identifiers

Chemical Name or Material Ethyl 2-cyanoacetate
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H7NO2
Linear Formula NCCH2CO2C2H5
Formula Weight 113.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB00844EE

  SDS
fisher scientific company
SB00844EE
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 113.12
Quantity 50g
Assay 97%
Density 1.0600g/mL
CAS 105-56-6
Assay Percent Range 97%
Molecular Formula C5H7NO2
Chemical Name or Material Ethyl 2-cyanoacetate
Linear Formula NCCH2CO2C2H5