Please login to your online account to display your discounted pricing

ethyl 2-cyanoacetate, Maybridge


Quantity 50g

Manufacturer: fisher scientific company  SB00844EE

 View more versions of this product

Catalog No. SB00844EE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 113.12
Quantity 50g
Assay 97%
Density 1.0600g/mL
CAS 105-56-6
Assay Percent Range 97%
Molecular Formula C5H7NO2
Chemical Name or Material Ethyl 2-cyanoacetate
Linear Formula NCCH2CO2C2H5

Specifications

Chemical Name or Material Ethyl 2-cyanoacetate
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H7NO2
Linear Formula NCCH2CO2C2H5
Formula Weight 113.12