missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 2-cyano-2-methylpropionate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3143218
Description
It is employed as a intermediates for pharmaceutical and organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a intermediates for pharmaceutical and organic synthesis.
Solubility
Soluble in ether. Not miscible or difficult to mix with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Ethyl 2-cyano-2-methylpropionate | |
0.97 | |
C7H11NO2 | |
50 g | |
Soluble in ether. Not miscible or difficult to mix with water. | |
CCOC(=O)C(C)(C)C#N | |
141.17 | |
141.17 |
1572-98-1 | |
185°C | |
MFCD00034667 | |
2-cyano-2-methylpropionic acid ethyl ester, ethyl 2-cyano-2-methylpropionate, ethyl 2-cyano-2,2-dimethylacetate, 2-cyano-2-methylpropanoic acid ethyl ester, ethyl 2-cyano-2-methyl-propanoate, propanoic acid, 2-cyano-2-methyl-, ethyl ester, pubchem23352, acmc-209dfv, ksc529g3h, ethyl2-cyano-2-methylpropionate | |
FYGRPGOHQCPZCV-UHFFFAOYSA-N | |
ethyl 2-cyano-2-methylpropanoate | |
344887 | |
97% |
Chemical Identifiers
1572-98-1 | |
141.17 | |
FYGRPGOHQCPZCV-UHFFFAOYSA-N | |
ethyl 2-cyano-2-methylpropanoate |
C7H11NO2 | |
MFCD00034667 | |
344887 | |
CCOC(=O)C(C)(C)C#N |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
H302-H318
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only