missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-chloropyrimidine-5-carboxylate, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC454140010

 View more versions of this product

Catalog No. AC454140010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Ethyl 2-chloropyrimidine-5-carboxylate
57°C to 58°C
89793-12-4
C7H7ClN2O2
IEMKQRSOAOPKRJ-UHFFFAOYSA-N
ethyl 2-chloropyrimidine-5-carboxylate
10487815
95%
Glass Bottle
1g
94% min. (GC)
2-chloro-5-ethoxycarbonyl pyrimidine, 2-chloro-5-pyrimidinecarboxylic acid ethyl ester, 2-chloro-pyrimidine-5-carboxylic acid ethyl ester, 5-pyrimidinecarboxylic acid, 2-chloro-, ethyl ester, acmc-209r2w, ethyl 2-chloranylpyrimidine-5-carboxylate, ethyl 2-chloro-pyrimidine-5-carboxylate, ethyl-2-chloropyrimidine-5-carboxylate, ethyl2-chloropyrimidine-5-carboxylate, ksc663i8r
CCOC(=O)C1=CN=C(N=C1)Cl
186.6
186.6

Chemical Identifiers

89793-12-4
186.6
2-chloro-5-ethoxycarbonyl pyrimidine, 2-chloro-5-pyrimidinecarboxylic acid ethyl ester, 2-chloro-pyrimidine-5-carboxylic acid ethyl ester, 5-pyrimidinecarboxylic acid, 2-chloro-, ethyl ester, acmc-209r2w, ethyl 2-chloranylpyrimidine-5-carboxylate, ethyl 2-chloro-pyrimidine-5-carboxylate, ethyl-2-chloropyrimidine-5-carboxylate, ethyl2-chloropyrimidine-5-carboxylate, ksc663i8r
ethyl 2-chloropyrimidine-5-carboxylate
C7H7ClN2O2
IEMKQRSOAOPKRJ-UHFFFAOYSA-N
10487815
CCOC(=O)C1=CN=C(N=C1)Cl
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water

GHS Signal Word: Warning

SDS Documents