Please login to your online account to display your discounted pricing

Ethyl 2-chloropyrimidine-5-carboxylate, 95%, Acros Organics


Quantity 250mg

Manufacturer: thermo fisher scientific  AC454142500

 View more versions of this product

Catalog No. AC454142500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Ethyl 2-chloropyrimidine-5-carboxylate
CAS 89793-12-4
Assay Percent Range 95%
Molecular Formula C7H7ClN2O2
Quantity 250mg
Formula Weight 186.6

Specifications

Chemical Name or Material Ethyl 2-chloropyrimidine-5-carboxylate
Assay Percent Range 95%
Molecular Formula C7H7ClN2O2
Formula Weight 186.6