Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-chloropropionate, 97%

Chemical Identifiers

Chemical Name or Material Ethyl 2-chloropropionate
CAS 535-13-7
Molecular Formula C5H9ClO2
Formula Weight 136.58
MDL Number MFCD00000864
UN Number UN2935
Beilstein 773906
Merck Index 14,3785
Synonym 2-Chloropropionic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1341018

  SDS
alfa aesar™
L1341018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1341030

  SDS
alfa aesar™
L1341030
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 136.58
Chemical Name or Material Ethyl 2-chloropropionate
Assay 97%
MDL Number MFCD00000864
Beilstein 773906
TSCA TSCA
Molecular Formula C5H9ClO2
CAS 535-13-7
UN Number UN2935
Merck Index 14,3785
Synonym 2-Chloropropionic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 146° to 149°C
Odor Pungent
Quantity 50g
Flash Point 38°C (100°F)
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYL 2-CHLOROPROPIONATE
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.