missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-Chloronicotinate, TCI America™

Manufacturer:  TCI America E0844100G

 View more versions of this product

Catalog No. E0844100G



Specifications

Specifications

Ethyl 2-Chloronicotinate
Yellow
1452-94-4
MFCD00674129
PMIMPBYTPPRBGD-UHFFFAOYSA-N
ethyl 2-chloropyridine-3-carboxylate
2796614
Clear Liquid at 20°C
78°C
100g
C8H8ClNO2
ethyl 2-chloronicotinate, 2-chloronicotinic acid ethyl ester, ethyl-2-chloronicotinate, 2-chloro-nicotinic acid ethyl ester, ethyl 2-chloro-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 2-chloro-, ethyl ester, 2-chloro-3-pyridinecarboxylic acid ethyl ester, ethyl 2chloronicotinate, acmc-1brbe, ethyl 2-chloro-nicotinate
CCOC(=O)C1=C(N=CC=C1)Cl
185.607
185.61
≥98.0% (GC)

Chemical Identifiers

1452-94-4
185.607
PMIMPBYTPPRBGD-UHFFFAOYSA-N
2796614
CCOC(=O)C1=C(N=CC=C1)Cl
C8H8ClNO2
MFCD00674129
ethyl 2-chloronicotinate, 2-chloronicotinic acid ethyl ester, ethyl-2-chloronicotinate, 2-chloro-nicotinic acid ethyl ester, ethyl 2-chloro-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 2-chloro-, ethyl ester, 2-chloro-3-pyridinecarboxylic acid ethyl ester, ethyl 2chloronicotinate, acmc-1brbe, ethyl 2-chloro-nicotinate
ethyl 2-chloropyridine-3-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

Documents