Please login to your online account to display your discounted pricing

Ethyl 2-chlorobenzoate 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC392981000

 View more versions of this product

Catalog No. AC392981000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 184.62
Quantity 100g
Color Clear colorless to pale yellow
Refractive Index 1.5220 to 1.5240 (20°C, 589nm)
Linear Formula ClC6H4C(O)OCH2CH3
Assay Percent Range 98%
Chemical Name or Material Ethyl 2-chlorobenzoate
MDL Number MFCD00016338
Packaging Glass bottle
Infrared Spectrum Authentic
CAS 7335-25-3
Molecular Formula C9 H9 Cl O2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material Ethyl 2-chlorobenzoate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9 H9 Cl O2
Linear Formula ClC6H4C(O)OCH2CH3
Formula Weight 184.62
MDL Number MFCD00016338
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents