Please login to your online account to display your discounted pricing

Ethyl 2-chlorobenzoate 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl 2-chlorobenzoate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9 H9 Cl O2
Linear Formula ClC6H4C(O)OCH2CH3
Formula Weight 184.62
MDL Number MFCD00016338
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC392981000

  SDS
thermo fisher scientific
AC392981000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 184.62
Quantity 100g
Color Clear colorless to pale yellow
Chemical Name or Material Ethyl 2-chlorobenzoate
Refractive Index 1.5220 to 1.5240 (20°C, 589nm)
Assay Percent Range 98%
Linear Formula ClC6H4C(O)OCH2CH3
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C9 H9 Cl O2
MDL Number MFCD00016338
CAS 7335-25-3
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.