missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 2-chlorobenzoate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1014136
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl 2-chlorobenzoate | |
1.194 | |
>110°C (230°F) | |
1.524 | |
500 g | |
2-chlorobenzoic acid ethyl ester, ethyl o-chlorobenzoate, benzoic acid, 2-chloro-, ethyl ester, ethyl-2-chlorobenzoate, ethyl2-chlorobenzoate, pubchem3703, acmc-1bfxj, ethyl 2-chloranylbenzoate, chlorobenzoic acid ethyl ester, ksc498a5r | |
CCOC(=O)C1=CC=CC=C1Cl | |
184.62 | |
184.62 |
7335-25-3 | |
243°C | |
C9H9ClO2 | |
MFCD00016338 | |
2207359 | |
RETLCWPMLJPOTP-UHFFFAOYSA-N | |
ethyl 2-chlorobenzoate | |
81783 | |
98% |
Chemical Identifiers
7335-25-3 | |
184.62 | |
RETLCWPMLJPOTP-UHFFFAOYSA-N | |
81783 | |
CCOC(=O)C1=CC=CC=C1Cl |
C9H9ClO2 | |
MFCD00016338 | |
2-chlorobenzoic acid ethyl ester, ethyl o-chlorobenzoate, benzoic acid, 2-chloro-, ethyl ester, ethyl-2-chlorobenzoate, ethyl2-chlorobenzoate, pubchem3703, acmc-1bfxj, ethyl 2-chloranylbenzoate, chlorobenzoic acid ethyl ester, ksc498a5r | |
ethyl 2-chlorobenzoate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 230-842-6
TSCA : Yes
RUO – Research Use Only