Learn More
Ethyl 2-chloroacetoacetate, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1981022
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl 2-chloroacetoacetate | |
<°C to 80°C | |
200°C | |
0.96 | |
1.4425 | |
100 g | |
ethyl 2-chloroacetoacetate, 2-chloroacetoacetic acid ethyl ester, butanoic acid, 2-chloro-3-oxo-, ethyl ester, ethyl alpha-chloroacetoacetate, ethyl-2-chloracetoacetate, 2-chloro-3-oxobutyric acid ethyl ester, ethyl2-chloroacetoacetate, 2-chloro-3-oxobutanoic acid, ethyl ester, ethyl2-chloro-3-oxobutanoate, ethyl 2-chloro-3-oxobutyrate | |
CCOC(=O)C(Cl)C(C)=O | |
164.59 | |
164.59 | |
Liquid |
609-15-4 | |
1.182 | |
54°C (180°F) | |
C6H9ClO3 | |
MFCD00009141 | |
774278 | |
RDULEYWUGKOCMR-UHFFFAOYNA-N | |
ethyl 2-chloro-3-oxobutanoate | |
11858 | |
96% |
Chemical Identifiers
609-15-4 | |
164.59 | |
RDULEYWUGKOCMR-UHFFFAOYNA-N | |
ethyl 2-chloro-3-oxobutanoate |
C6H9ClO3 | |
MFCD00009141 | |
11858 | |
CCOC(=O)C(Cl)C(C)=O |
Safety and Handling
GHS H Statement
H226-H302-H315-H319-H335
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302-H315-H319-H335
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 210-180-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only