Please login to your online account to display your discounted pricing

Ethyl 2-chloroacetoacetate, 90%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl 2-chloroacetoacetate
Assay ≥88%
Assay Percent Range 90%
Molecular Formula C6H9ClO3
Linear Formula CH3COCH(Cl)CO2C2H5
Formula Weight 164.59
MDL Number MFCD00009141
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC376312500

  SDS
thermo fisher scientific
AC376312500
Glass bottle 250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC376310010

  SDS
thermo fisher scientific
AC376310010
Glass bottle 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥88%
Physical Form Liquid
Formula Weight 164.59
Quantity 250mL
Color Clear yellow
Density 1.1900g/mL
Packaging Glass bottle
Linear Formula CH3COCH(Cl)CO2C2H5
Chemical Name or Material Ethyl 2-chloroacetoacetate
Infrared Spectrum Authentic
Refractive Index 1.4410 to 1.4440 (20°C, 589nm)
Assay Percent Range 90%
Molecular Formula C6H9ClO3
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00009141
CAS 609-15-4
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.