Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-chloro-6-methylpyridine-4-carboxylate, 97%

$70.95 - $480.26

Chemical Identifiers

Chemical Name or Material Ethyl 2-chloro-6-methylpyridine-4-carboxylate
CAS 3998-88-7
Molecular Formula C9H10ClNO2
Formula Weight 199.64
MDL Number MFCD09842151
Synonym 2-Chloro-6-methylpyridine-4-carboxylic acid ethyl ester; Ethyl 2-chloro-6-methylisonicotinate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2777803

  SDS
alfa aesar™
H2777803
1g Each for $136.69

AAH2777806

  SDS
alfa aesar™
H2777806
5g Each for $480.26

AAH27778MD

  SDS
alfa aesar™
H27778MD
250mg Each for $70.95
Description & Specifications

Specifications

Formula Weight 199.64
Chemical Name or Material Ethyl 2-chloro-6-methylpyridine-4-carboxylate
Assay 97%
Molecular Formula C9H10ClNO2
MDL Number MFCD09842151
CAS 3998-88-7
Synonym 2-Chloro-6-methylpyridine-4-carboxylic acid ethyl ester; Ethyl 2-chloro-6-methylisonicotinate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 107°C (1.5mmHg)
Melting Point 65° to 67°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.