Please login to your online account to display your discounted pricing

Ethyl 2-chloro-4-methyl-1,3-thiazole-5-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Ethyl 2-chloro-4-methyl-1, 3-thiazole-5-carboxylate
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H8ClNO2S
Formula Weight 205.66
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO07617CB

  SDS
fisher scientific company
MO07617CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MO07617DA

  SDS
fisher scientific company
MO07617DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 205.66
Quantity 250mg
Assay 97%
Chemical Name or Material Ethyl 2-chloro-4-methyl-1, 3-thiazole-5-carboxylate
CAS 7238-62-2
Molecular Formula C7H8ClNO2S
Assay Percent Range 97%