Please login to your online account to display your discounted pricing

Ethyl 2-chloro-4-methyl-1,3-thiazole-5-carboxylate, 97%, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  MO07617DA

 View more versions of this product

Catalog No. MO07617DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 205.66
Quantity 1g
Assay 97%
Chemical Name or Material Ethyl 2-chloro-4-methyl-1, 3-thiazole-5-carboxylate
Assay Percent Range 97%
Molecular Formula C7H8ClNO2S
CAS 7238-62-2

Specifications

Chemical Name or Material Ethyl 2-chloro-4-methyl-1, 3-thiazole-5-carboxylate
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H8ClNO2S
Formula Weight 205.66