missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-chloro-4-iodonicotinate, 97%

$247.82 - $1,097.50

Chemical Identifiers

CAS 219727-21-6
Molecular Formula C8H7ClINO2
Formula Weight 311.51
MDL Number MFCD07781133
Synonym 2-Chloro-4-iodonicotinic acid ethyl ester; Ethyl 2-chloro-4-iodopyridine-3-carboxylate
Chemical Name or Material Ethyl 2-chloro-4-iodonicotinate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5008503 SDS Alfa Aesar™
H5008503
1g Each for $247.82
AAH5008506 SDS Alfa Aesar™
H5008506
5g Each for $1,097.50
Specifications Description & Specifications

Chemical Identifiers

219727-21-6
311.51
2-Chloro-4-iodonicotinic acid ethyl ester; Ethyl 2-chloro-4-iodopyridine-3-carboxylate
C8H7ClINO2
MFCD07781133
Ethyl 2-chloro-4-iodonicotinate

Specifications

219727-21-6
311.51
2-Chloro-4-iodonicotinic acid ethyl ester; Ethyl 2-chloro-4-iodopyridine-3-carboxylate
Ethyl 2-chloro-4-iodonicotinate
C8H7ClINO2
MFCD07781133
1g
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning