Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-chloro-4,4,4-trifluoroacetoacetate, 94%

Chemical Identifiers

Chemical Name or Material Ethyl 2-chloro-4,4,4-trifluoroacetoacetate
CAS 363-58-6
Molecular Formula C6H6ClF3O3
Formula Weight 218.56
MDL Number MFCD00041540
UN Number UN1760
Beilstein 1787023
Synonym Ethyl 2-chloro-4,4,4-trifluoro-3-oxobutyrate; 2-Chloro-4,4,4-trifluoroacetoacetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2227006

  SDS
alfa aesar™
B2227006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2227014

  SDS
alfa aesar™
B2227014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 218.56
Chemical Name or Material Ethyl 2-chloro-4,4,4-trifluoroacetoacetate
Assay 94%
Molecular Formula C6H6ClF3O3
MDL Number MFCD00041540
CAS 363-58-6
UN Number UN1760
Beilstein 1787023
Synonym Ethyl 2-chloro-4,4,4-trifluoro-3-oxobutyrate; 2-Chloro-4,4,4-trifluoroacetoacetic acid ethyl ester
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 67° to 71°C (35mmHg)
Quantity 5g
Flash Point 67°C (153°F)
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.