Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-chloro-3-pyridylglyoxylate, 97%

$269.72 - $1,193.35

Chemical Identifiers

Chemical Name or Material Ethyl 2-chloro-3-pyridylglyoxylate
CAS 902837-56-3
Molecular Formula C9H8ClNO3
Formula Weight 213.62
MDL Number MFCD07781194
Synonym 2-Chloro-3-pyridylglyoxylic acid ethyl ester; Ethyl (2-chloro-3-pyridinyl)oxoacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5015303

  SDS
alfa aesar™
H5015303
1g Each for $269.72

AAH5015306

  SDS
alfa aesar™
H5015306
5g Each for $1,193.35
Description & Specifications

Specifications

Formula Weight 213.62
Quantity 1g
Chemical Name or Material Ethyl 2-chloro-3-pyridylglyoxylate
Assay 97%
Molecular Formula C9H8ClNO3
MDL Number MFCD07781194
CAS 902837-56-3
Synonym 2-Chloro-3-pyridylglyoxylic acid ethyl ester; Ethyl (2-chloro-3-pyridinyl)oxoacetate

Applications
Ethyl 2-chloro-3-pyridylglyoxylate is used as a pharmaceutical intermediates.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.