missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-Butylacetoacetate, TCI America™

$73.33 - $220.00

Chemical Identifiers

CAS 1540-29-0
Molecular Formula C10H18O3
Molecular Weight (g/mol) 186.251
MDL Number MFCD00026888
InChI Key ZTOQBHVLCJERBS-UHFFFAOYSA-N
Synonym ethyl 2-n-butylacetoacetate, ethyl 2-butylacetoacetate, hexanoic acid, 2-acetyl-, ethyl ester, ethyl .alpha.-butylacetoacetate, 2-acetylhexanoic acid ethyl ester, ethyl butyl acetoacetate, ethyl 2-acetyl hexanoate, acmc-209da5, ethyl-2-n-butylacetoacetate, ksc494i1n
PubChem CID 95466
IUPAC Name ethyl 2-acetylhexanoate
SMILES CCCCC(C(=O)C)C(=O)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B07025ML View Documents TCI America
B07025ML
5mL Each for $73.33
 
B070225ML View Documents TCI America
B070225ML
25mL Each for $220.00
 
Specifications

Chemical Identifiers

1540-29-0
186.251
ZTOQBHVLCJERBS-UHFFFAOYSA-N
95466
CCCCC(C(=O)C)C(=O)OCC
C10H18O3
MFCD00026888
ethyl 2-n-butylacetoacetate, ethyl 2-butylacetoacetate, hexanoic acid, 2-acetyl-, ethyl ester, ethyl .alpha.-butylacetoacetate, 2-acetylhexanoic acid ethyl ester, ethyl butyl acetoacetate, ethyl 2-acetyl hexanoate, acmc-209da5, ethyl-2-n-butylacetoacetate, ksc494i1n
ethyl 2-acetylhexanoate

Specifications

224°C
5mL
C10H18O3
ethyl 2-n-butylacetoacetate, ethyl 2-butylacetoacetate, hexanoic acid, 2-acetyl-, ethyl ester, ethyl .alpha.-butylacetoacetate, 2-acetylhexanoic acid ethyl ester, ethyl butyl acetoacetate, ethyl 2-acetyl hexanoate, acmc-209da5, ethyl-2-n-butylacetoacetate, ksc494i1n
CCCCC(C(=O)C)C(=O)OCC
186.251
186.25
≥96.0% (GC)
Colorless
1540-29-0
MFCD00026888
ZTOQBHVLCJERBS-UHFFFAOYSA-N
ethyl 2-acetylhexanoate
95466
Clear Liquid at 20°C
Ethyl 2-Butylacetoacetate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1505

TSCA : No