missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 2-Bromovalerate, TCI America™

Manufacturer:  TCI America E097025G

 View more versions of this product

Catalog No. E097025G



Specifications

Specifications

Ethyl 2-Bromovalerate
Yellow
615-83-8
MFCD00000159
ethyl 2-bromovalerate, ethyl2-bromovalerate, ethyl .alpha.-bromovalerate, pentanoic acid, 2-bromo-, ethyl ester, 2-bromovaleric acid ethyl ester, 2-bromopentanoic acid ethyl ester, 2-bromo-pentanoic acid ethyl ester, pentanoic acid, bromo-, ethyl ester, ethyl bromovalerate, ethyl-2bromovalerate
CCCC(C(=O)OCC)Br
209.083
209.08
≥98.0% (GC)
94°C
25g
C7H13BrO2
3265
ORSIRXYHFPHWTN-UHFFFAOYSA-N
ethyl 2-bromopentanoate
79071
Clear Liquid at 20°C

Chemical Identifiers

615-83-8
209.083
ORSIRXYHFPHWTN-UHFFFAOYSA-N
79071
CCCC(C(=O)OCC)Br
C7H13BrO2
MFCD00000159
ethyl 2-bromovalerate, ethyl2-bromovalerate, ethyl .alpha.-bromovalerate, pentanoic acid, 2-bromo-, ethyl ester, 2-bromovaleric acid ethyl ester, 2-bromopentanoic acid ethyl ester, 2-bromo-pentanoic acid ethyl ester, pentanoic acid, bromo-, ethyl ester, ethyl bromovalerate, ethyl-2bromovalerate
ethyl 2-bromopentanoate
Safety and Handling

Safety and Handling

TSCA : No

Documents