missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-bromothiazole-5-carboxylate, 98%

Manufacturer:  Alfa Aesar™ H5510906

 View more versions of this product

Catalog No. AAH5510906



Description

Description

Applications
Ethyl 2-bromothiazole-5-carboxylate is used in medicine, pharmaceutical intermediate

Solubility
Miscible is water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Ethyl 2-bromothiazole-5-carboxylate
216°C to 217°C
41731-83-3
MFCD00463837
KTYIFXLNIMPSKI-UHFFFAOYSA-N
ethyl 2-bromo-1,3-thiazole-5-carboxylate
3614103
98%
110°C (230°F)
5g
C6H6BrNO2S
2-bromo-5-ethoxycarbonyl thiazole
CCOC(=O)C1=CN=C(S1)Br
236.083
236.08

Chemical Identifiers

41731-83-3
236.083
KTYIFXLNIMPSKI-UHFFFAOYSA-N
3614103
CCOC(=O)C1=CN=C(S1)Br
C6H6BrNO2S
MFCD00463837
2-bromo-5-ethoxycarbonyl thiazole
ethyl 2-bromo-1,3-thiazole-5-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H317
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents