Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-bromooctanoate, 98+%

$60.97 - $174.53

Chemical Identifiers

Chemical Name or Material Ethyl 2-bromooctanoate
CAS 5445-29-4
Molecular Formula C10H19BrO2
Formula Weight 251.17g/mol
MDL Number MFCD00042603
Beilstein 1767535
Synonym 2-Bromooctanoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2189614

 
alfa aesar™
B21896-14
25g Each for $60.97

AAB2189622

 
alfa aesar™
B21896-22
100g Each for $174.53
Description & Specifications

Specifications

Boiling Point 137° to 140°C (25mmHg)
Quantity 25g
Flash Point 106°C (222°F)
Chemical Name or Material Ethyl 2-bromooctanoate
CAS 5445-29-4
Assay ≥98%
Molecular Formula C10H19BrO2
Formula Weight 251.17g/mol
MDL Number MFCD00042603
Beilstein 1767535
Synonym 2-Bromooctanoic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.