missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 2-bromoisobutyrate, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 118135000
Specifications
Ethyl 2-bromoisobutyrate | |
60°C | |
Glass bottle | |
1.329 | |
Colorless to Yellow | |
98% | |
97.5% min. (GC) | |
(CH3)2C(Br)CO2C2H5 | |
02, 296 | |
IOLQWGVDEFWYNP-UHFFFAOYSA-N | |
ethyl 2-bromo-2-methylpropanoate | |
11745 | |
Liquid |
1.3290g/mL | |
Authentic | |
1.4430 to 1.446 | |
161°C to 163°C | |
500g | |
600-00-0 | |
C6H11BrO2 | |
MFCD00000123 | |
2-bromo-2-methylpropanoic acid, ethyl ester, ethyl .alpha.-bromoisobutyrate, ethyl 2-bromo-2-methyl-propanoate, ethyl 2-bromo-2-methylpropionate, ethyl 2-bromoisobutyrate, ethyl alpha-bromoisobutyrate, ethyl-2-bromoisobutyrate, propanoic acid, 2-bromo-2-methyl-, ethyl ester, propanoic acid, bromo-2-methyl-, ethyl ester, propionic acid, 2-bromo-2-methyl-, ethyl ester | |
CCOC(=O)C(C)(C)Br | |
195.06 | |
195.06 | |
98% |
Chemical Identifiers
600-00-0 | |
195.06 | |
IOLQWGVDEFWYNP-UHFFFAOYSA-N | |
11745 | |
CCOC(=O)C(C)(C)Br |
C6H11BrO2 | |
MFCD00000123 | |
2-bromo-2-methylpropanoic acid, ethyl ester, ethyl .alpha.-bromoisobutyrate, ethyl 2-bromo-2-methyl-propanoate, ethyl 2-bromo-2-methylpropionate, ethyl 2-bromoisobutyrate, ethyl alpha-bromoisobutyrate, ethyl-2-bromoisobutyrate, propanoic acid, 2-bromo-2-methyl-, ethyl ester, propanoic acid, bromo-2-methyl-, ethyl ester, propionic acid, 2-bromo-2-methyl-, ethyl ester | |
ethyl 2-bromo-2-methylpropanoate |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause genetic defects if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Danger
EINECSNumber : 209-980-6
TSCA : TSCA