Learn More
Ethyl 2-bromoisobutyrate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 118130050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ethyl 2-bromoisobutyrate | |
600-00-0 | |
161°C to 163°C | |
Authentic | |
Glass bottle | |
1.4430 to 1.446 | |
MFCD00000123 | |
02, 296 | |
ethyl 2-bromoisobutyrate, ethyl 2-bromo-2-methylpropionate, ethyl alpha-bromoisobutyrate, ethyl .alpha.-bromoisobutyrate, propanoic acid, 2-bromo-2-methyl-, ethyl ester, 2-bromo-2-methylpropanoic acid, ethyl ester, ethyl-2-bromoisobutyrate, propanoic acid, bromo-2-methyl-, ethyl ester, ethyl 2-bromo-2-methyl-propanoate, propionic acid, 2-bromo-2-methyl-, ethyl ester | |
CCOC(=O)C(C)(C)Br | |
195.06 | |
195.06 |
98% | |
1.3290g/mL | |
60°C | |
97.5% min. (GC) | |
C6H11BrO2 | |
(CH3)2C(Br)CO2C2H5 | |
5g | |
1.329 | |
IOLQWGVDEFWYNP-UHFFFAOYSA-N | |
ethyl 2-bromo-2-methylpropanoate | |
11745 | |
98% |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause genetic defects if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Danger
EINECSNumber : 209-980-6
TSCA : TSCA
RUO â Research Use Only