Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-bromobutyrate, 97%

Chemical Identifiers

Chemical Name or Material Ethyl 2-bromobutyrate
CAS 533-68-6
Molecular Formula C6H11BrO2
Formula Weight 195.06
MDL Number MFCD00000155
UN Number UN3272
Beilstein 969841
Merck Index 14,1413
Synonym 2-Bromobutyric acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1272906

 
alfa aesar™
A1272906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1272922

 
alfa aesar™
A1272922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1272930

 
alfa aesar™
A1272930
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 195.06
Chemical Name or Material Ethyl 2-bromobutyrate
TSCA TSCA
Assay 97%
Melting Point −4°C
Boiling Point 176° to 178°C
Molecular Formula C6H11BrO2
Quantity 5g
Flash Point 58°C (136°F)
MDL Number MFCD00000155
CAS 533-68-6
UN Number UN3272
Beilstein 969841
Merck Index 14,1413
Synonym 2-Bromobutyric acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

Applications
Ethyl 2-bromobutyrate, is an important raw material and an intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used for agrochemical and pharmaceutical intermediate.

Solubility
Insoluble in water. Miscible with alcohol and ether

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: 2 - 8°C. Light sensitive. Keep away from strong oxidizing agents, acids, bases, oxidizing agents, reducing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.