Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-bromobenzoate, 98+%

$38.12 - $476.14

Chemical Identifiers

Chemical Name or Material Ethyl 2-bromobenzoate
CAS 6091-64-1
Molecular Formula C9H9BrO2
Formula Weight 229.07
MDL Number MFCD00015443
Beilstein 2207360
Synonym 2-Bromobenzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1316609

  SDS
alfa aesar™
A1316609
10g Each for $38.12

AAA1316618

  SDS
alfa aesar™
A1316618
50g Each for $106.53

AAA1316630

  SDS
alfa aesar™
A1316630
250g Each for $476.14
Description & Specifications

Specifications

Formula Weight 229.07
Chemical Name or Material Ethyl 2-bromobenzoate
Assay ≥98%
Boiling Point 255°C
Molecular Formula C9H9BrO2
Quantity 10g
Flash Point >110°C (230°F)
MDL Number MFCD00015443
CAS 6091-64-1
Beilstein 2207360
Synonym 2-Bromobenzoic acid ethyl ester