missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-bromobenzenesulfonate, 97%

Chemical Identifiers

CAS 99769-29-6
Molecular Formula C8H9BrO3S
Molecular Weight (g/mol) 265.121
MDL Number MFCD22683123
InChI Key QUZFLGXJHUNSEH-UHFFFAOYSA-N
Synonym 2-Bromobenzenesulfonic acid ethyl ester
PubChem CID 97030407
IUPAC Name ethyl 2-bromobenzenesulfonate
SMILES CCOS(=O)(=O)C1=CC=CC=C1Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60402MD View Documents Alfa Aesar™
H60402MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6040203 View Documents Alfa Aesar™
H6040203
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

99769-29-6
265.121
QUZFLGXJHUNSEH-UHFFFAOYSA-N
97030407
CCOS(=O)(=O)C1=CC=CC=C1Br
C8H9BrO3S
MFCD22683123
2-Bromobenzenesulfonic acid ethyl ester
ethyl 2-bromobenzenesulfonate

Specifications

250mg
C8H9BrO3S
2-Bromobenzenesulfonic acid ethyl ester
CCOS(=O)(=O)C1=CC=CC=C1Br
265.121
265.12
Ethyl 2-bromobenzenesulfonate
99769-29-6
MFCD22683123
QUZFLGXJHUNSEH-UHFFFAOYSA-N
ethyl 2-bromobenzenesulfonate
97030407
97%