Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-bromo-4-methylthiazole-5-carboxylate, 97%

$66.61 - $247.11

Chemical Identifiers

Chemical Name or Material Ethyl 2-bromo-4-methylthiazole-5-carboxylate
CAS 22900-83-0
Molecular Formula C7H8BrNO2S
Formula Weight 250.12
MDL Number MFCD03791227
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5183303

  SDS
alfa aesar™
H5183303
1g Each for $66.61

AAH5183306

  SDS
alfa aesar™
H5183306
5g Each for $247.11
Description & Specifications

Specifications

Formula Weight 250.12
Molecular Formula C7H8BrNO2S
MDL Number MFCD03791227
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material Ethyl 2-bromo-4-methylthiazole-5-carboxylate
Quantity 1g
Assay 97%
CAS 22900-83-0

Applications
It finds its application in the preparation of thiazole-4- and -5-carboxylates, and an infrared study of their rotational isomers and in the discovery of thiazolylpyridinone SCD1 inhibitors with preferential liver distribution and reduced mechanism-based adverse effects.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.