Learn More
Ethyl 2-bromo-4,4,4-trifluorobutyrate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H32572MD
Description
Ethyl 2-bromo-4,4,4-trifluorobutyrate is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 2-bromo-4,4,4-trifluorobutyrate is used as a pharmaceutical intermediate.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Ethyl 2-bromo-4,4,4-trifluorobutyrate | |
67°C (18 mmHg) | |
MFCD10687235 | |
UN3265 | |
POVYUFTVLGOXEE-UHFFFAOYSA-N | |
ethyl 2-bromo-4,4,4-trifluorobutanoate | |
45594306 | |
97% |
367-33-9 | |
C6H8BrF3O2 | |
250 mg | |
2-bromo-4,4,4-trifluorobutyric acid ethyl ester, ethyl 2-bromo-4,4,4-trifluorobutyrate, ethyl 2-bromo-4,4,4-trifluoro-butanoate, butanoic acid,2-bromo-4,4,4-trifluoro-, ethyl ester | |
CCOC(=O)C(CC(F)(F)F)Br | |
249.027 | |
249.03 |
Chemical Identifiers
367-33-9 | |
249.027 | |
POVYUFTVLGOXEE-UHFFFAOYSA-N | |
45594306 | |
CCOC(=O)C(CC(F)(F)F)Br |
C6H8BrF3O2 | |
MFCD10687235 | |
2-bromo-4,4,4-trifluorobutyric acid ethyl ester, ethyl 2-bromo-4,4,4-trifluorobutyrate, ethyl 2-bromo-4,4,4-trifluoro-butanoate, butanoic acid,2-bromo-4,4,4-trifluoro-, ethyl ester | |
ethyl 2-bromo-4,4,4-trifluorobutanoate |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only