Please login to your online account to display your discounted pricing

Ethyl 2-benzylacetoacetate 97%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  118101000

 View more versions of this product

Catalog No. AC118101000

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Ethyl 2-benzylacetoacetate
Name Note 97%
CAS 620-79-1
Density 1.03g/mL
EINECS Number 210-651-4
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4986 to 1.5006 (20°C, 589nm)
Physical Form Liquid
Assay ≥96.0%
Assay Percent Range 97%
Color Clear light yellow
Molecular Formula C13H16O3
Quantity 100g
Linear Formula CH3COCH(CH2C6H5)CO2C2H5
Formula Weight 220.27
MDL Number MFCD00009156
Beilstein 10, 710

Specifications

Chemical Name or Material Ethyl 2-benzylacetoacetate
Name Note 97%
CAS 620-79-1
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C13H16O3
Linear Formula CH3COCH(CH2C6H5)CO2C2H5
Formula Weight 220.27
MDL Number MFCD00009156