Please login to your online account to display your discounted pricing

Ethyl 2-benzylacetoacetate 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl 2-benzylacetoacetate
Name Note 97%
CAS 620-79-1
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C13H16O3
Linear Formula CH3COCH(CH2C6H5)CO2C2H5
Formula Weight 220.27
MDL Number MFCD00009156
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC118101000

  SDS
thermo fisher scientific
118101000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 220.27
Quantity 100g
Color Clear light yellow
EINECS Number 210-651-4
Density 1.03g/mL
Beilstein 10, 710
Infrared Spectrum Authentic
Refractive Index 1.4986 to 1.5006 (20°C, 589nm)
Packaging Glass bottle
Assay Percent Range 97%
Name Note 97%
Molecular Formula C13H16O3
Linear Formula CH3COCH(CH2C6H5)CO2C2H5
Chemical Name or Material Ethyl 2-benzylacetoacetate
MDL Number MFCD00009156
CAS 620-79-1