missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-benzylacetoacetate, 97%

$90.22 - $226.69

Chemical Identifiers

CAS 620-79-1
Molecular Formula C13H16O3
Molecular Weight (g/mol) 220.268
MDL Number MFCD00009156
InChI Key XDWQYMXQMNUWID-UHFFFAOYSA-N
Synonym ethyl 2-benzylacetoacetate, ethyl 2-acetyl-3-phenylpropionate, ethyl benzyl acetoacetate, ethyl alpha-benzylacetoacetate, 2-benzylacetoacetic acid ethyl ester, ethyl .alpha.-benzylacetoacetate, 2-benzylacetoacetate, ethyl 3-oxo-2-benzylbutanoate, ethyl alpha-acetylhydrocinnamate, ethyl benzylacetoacetate
PubChem CID 246929
IUPAC Name ethyl 2-benzyl-3-oxobutanoate
SMILES CCOC(=O)C(CC1=CC=CC=C1)C(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0157314 View Documents Alfa Aesar™
L0157314
25g Each for $90.22
 
AAL0157322 View Documents Alfa Aesar™
L0157322
100g Each for $226.69
 
Description

Description

Applications
Ethyl 2-benzylacetoacetate is used as a flavoring agent.

Solubility
Difficult in mix.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Chemical Identifiers

620-79-1
220.268
XDWQYMXQMNUWID-UHFFFAOYSA-N
246929
CCOC(=O)C(CC1=CC=CC=C1)C(=O)C
C13H16O3
MFCD00009156
ethyl 2-benzylacetoacetate, ethyl 2-acetyl-3-phenylpropionate, ethyl benzyl acetoacetate, ethyl alpha-benzylacetoacetate, 2-benzylacetoacetic acid ethyl ester, ethyl .alpha.-benzylacetoacetate, 2-benzylacetoacetate, ethyl 3-oxo-2-benzylbutanoate, ethyl alpha-acetylhydrocinnamate, ethyl benzylacetoacetate
ethyl 2-benzyl-3-oxobutanoate

Specifications

>110°C (230°F)
25g
C13H16O3
520909
XDWQYMXQMNUWID-UHFFFAOYSA-N
ethyl 2-benzyl-3-oxobutanoate
246929
97%
275°C to 276°C
620-79-1
MFCD00009156
ethyl 2-benzylacetoacetate, ethyl 2-acetyl-3-phenylpropionate, ethyl benzyl acetoacetate, ethyl alpha-benzylacetoacetate, 2-benzylacetoacetic acid ethyl ester, ethyl .alpha.-benzylacetoacetate, 2-benzylacetoacetate, ethyl 3-oxo-2-benzylbutanoate, ethyl alpha-acetylhydrocinnamate, ethyl benzylacetoacetate
CCOC(=O)C(CC1=CC=CC=C1)C(=O)C
220.268
220.27
Ethyl 2-benzylacetoacetate