Learn More
Ethyl 2-aminothiophene-3-carboxylate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6155914
Description
Ethyl 2-aminothiophene-3-carboxylate is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 2-aminothiophene-3-carboxylate is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Ethyl 2-aminothiophene-3-carboxylate | |
C7H9NO2S | |
25 g | |
Slightly soluble in water. | |
CCOC(=O)C1=C(SC=C1)N | |
171.214 | |
171.22 | |
Solid |
31891-06-2 | |
MFCD01566303 | |
2-amino-thiophene-3-carboxylic acid ethyl ester, 2-aminothiophene-3-carboxylic acid ethyl ester, ethyl2-aminothiophene-3-carboxylate, 2-amino-3-ethoxycarbonylthiophene, ethyl 2-amino-3-thiophenecarboxylate, 2-aminothiophene-3-carboxylate, 3-thiophenecarboxylic acid, 2-amino-, ethyl ester, pubchem22914, acmc-209hpg, ethyl 2-amino-3-thenoate | |
MKJQYFVTEPGXIE-UHFFFAOYSA-N | |
ethyl 2-aminothiophene-3-carboxylate | |
1988156 | |
98% |
Chemical Identifiers
31891-06-2 | |
171.214 | |
MKJQYFVTEPGXIE-UHFFFAOYSA-N | |
1988156 | |
CCOC(=O)C1=C(SC=C1)N |
C7H9NO2S | |
MFCD01566303 | |
2-amino-thiophene-3-carboxylic acid ethyl ester, 2-aminothiophene-3-carboxylic acid ethyl ester, ethyl2-aminothiophene-3-carboxylate, 2-amino-3-ethoxycarbonylthiophene, ethyl 2-amino-3-thiophenecarboxylate, 2-aminothiophene-3-carboxylate, 3-thiophenecarboxylic acid, 2-amino-, ethyl ester, pubchem22914, acmc-209hpg, ethyl 2-amino-3-thenoate | |
ethyl 2-aminothiophene-3-carboxylate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only