missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-aminothiazole-4-acetate, 97%

$24.91 - $355.19

Chemical Identifiers

CAS 53266-94-7
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.229
MDL Number MFCD00005330
InChI Key SHQNGLYXRFCPGZ-UHFFFAOYSA-N
Synonym 2-amino-4-thiazolyl acetic acid ethyl ester
PubChem CID 104454
IUPAC Name ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate
SMILES CCOC(=O)CC1=CSC(=N1)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54717MD View Documents Alfa Aesar™
H54717MD
250mg Each for $24.91
 
AAH5471703 View Documents Alfa Aesar™
H5471703
1g Each for $78.09
 
AAH5471706 View Documents Alfa Aesar™
H5471706
5g Each for $355.19
 
Specifications

Chemical Identifiers

53266-94-7
186.229
SHQNGLYXRFCPGZ-UHFFFAOYSA-N
104454
CCOC(=O)CC1=CSC(=N1)N
C7H10N2O2S
MFCD00005330
2-amino-4-thiazolyl acetic acid ethyl ester
ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate

Specifications

250mg
C7H10N2O2S
2-amino-4-thiazolyl acetic acid ethyl ester
CCOC(=O)CC1=CSC(=N1)N
186.229
186.23
Ethyl 2-aminothiazole-4-acetate
53266-94-7
MFCD00005330
SHQNGLYXRFCPGZ-UHFFFAOYSA-N
ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate
104454
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 258-452-1

TSCA : No

Recommended Storage : Ambient temperatures