Learn More
Ethyl 2-aminothiazole-4-acetate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H5471703
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ethyl 2-aminothiazole-4-acetate | |
C7H10N2O2S | |
1 g | |
SHQNGLYXRFCPGZ-UHFFFAOYSA-N | |
ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate | |
104454 | |
97% |
53266-94-7 | |
MFCD00005330 | |
ethyl 2-amino-4-thiazoleacetate, ethyl 2-2-aminothiazol-4-yl acetate, ethyl 2-2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazole-4-acetate, ethyl 2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazol-4-acetate, 2-amino-4-thiazolyl acetic acid ethyl ester, 4-thiazoleacetic acid, 2-amino-, ethyl ester, ethyl 2-amino-4-thiazolyl acetate | |
CCOC(=O)CC1=CSC(N)=N1 | |
186.23 | |
186.23 |
Chemical Identifiers
53266-94-7 | |
186.23 | |
SHQNGLYXRFCPGZ-UHFFFAOYSA-N | |
104454 | |
CCOC(=O)CC1=CSC(N)=N1 |
C7H10N2O2S | |
MFCD00005330 | |
ethyl 2-amino-4-thiazoleacetate, ethyl 2-2-aminothiazol-4-yl acetate, ethyl 2-2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazole-4-acetate, ethyl 2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazol-4-acetate, 2-amino-4-thiazolyl acetic acid ethyl ester, 4-thiazoleacetic acid, 2-amino-, ethyl ester, ethyl 2-amino-4-thiazolyl acetate | |
ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 258-452-1
TSCA : No
Recommended Storage : Ambient temperatures