missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-aminopyridine-4-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H6128006

 View more versions of this product

Catalog No. AAH6128006



Description

Description

Applications
Ethyl 2-aminopyridine-4-carboxylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

Ethyl 2-aminopyridine-4-carboxylate
5g
C8H10N2O2
ethyl 2-aminoisonicotinate, 2-amino-isonicotinic acid ethyl ester, ethyl 2-amino-4-pyridinecarboxylate, 2-aminoisonicotinic acid ethyl ester, 2-amino-4-ethoxycarbonyl pyridine, 4-pyridinecarboxylic acid, 2-amino-, ethyl ester, 2-amino-4-pyridinecarboxylic acid ethyl ester, 2-aminopyridine-4-carboxylic acid ethyl ester, pubchem16599, acmc-209bt6
CCOC(=O)C1=CC(=NC=C1)N
166.18
166.18
97%
White
13362-30-6
MFCD03791260
XVBZFXZNJAFCHL-UHFFFAOYSA-N
ethyl 2-aminopyridine-4-carboxylate
7015521
Powder

Chemical Identifiers

13362-30-6
166.18
XVBZFXZNJAFCHL-UHFFFAOYSA-N
7015521
CCOC(=O)C1=CC(=NC=C1)N
C8H10N2O2
MFCD03791260
ethyl 2-aminoisonicotinate, 2-amino-isonicotinic acid ethyl ester, ethyl 2-amino-4-pyridinecarboxylate, 2-aminoisonicotinic acid ethyl ester, 2-amino-4-ethoxycarbonyl pyridine, 4-pyridinecarboxylic acid, 2-amino-, ethyl ester, 2-amino-4-pyridinecarboxylic acid ethyl ester, 2-aminopyridine-4-carboxylic acid ethyl ester, pubchem16599, acmc-209bt6
ethyl 2-aminopyridine-4-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents