missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-aminobenzothiazole-6-carboxylate, 97%

$47.61 - $779.78

Chemical Identifiers

CAS 50850-93-6
Molecular Formula C10H10N2O2S
Molecular Weight (g/mol) 222.262
MDL Number MFCD00102724
InChI Key VYJSGJXWKSDUSG-UHFFFAOYSA-N
Synonym 2-amino benzo thiazole-6-carboxylic acid ethyl ester, 2-amino-6-ethoxycarbonylbenzothiazole, 2-amino-benzothiazole-6-carboxylic acid ethyl ester, 2-aminobenzothiazole-6-carboxylic acid ethyl ester, 3kid, 6-benzothiazolecarboxylic acid, 2-amino-, ethyl ester, ethyl 2-amino-benzothiazole-6-carboxylate, ethyl 2-aminobenzo d thiazole-6-carboxylate, ethyl 2-aminobenzothiazole-6-carboxylate, ethyl2-amino-benzothiazole-6-carboxylate
PubChem CID 601008
IUPAC Name ethyl 2-amino-1,3-benzothiazole-6-carboxylate
SMILES CCOC(=O)C1=CC2=C(C=C1)N=C(S2)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6498803
View Documents
Alfa Aesar™
H6498803
1g Each for $47.61
 
AAH6498806
View Documents
Alfa Aesar™
H6498806
5g Each for $188.78
 
AAH6498814
View Documents
Alfa Aesar™
H6498814
25g Each for $779.78
 
Specifications

Chemical Identifiers

50850-93-6
222.262
VYJSGJXWKSDUSG-UHFFFAOYSA-N
601008
CCOC(=O)C1=CC2=C(C=C1)N=C(S2)N
C10H10N2O2S
MFCD00102724
2-amino benzo thiazole-6-carboxylic acid ethyl ester, 2-amino-6-ethoxycarbonylbenzothiazole, 2-amino-benzothiazole-6-carboxylic acid ethyl ester, 2-aminobenzothiazole-6-carboxylic acid ethyl ester, 3kid, 6-benzothiazolecarboxylic acid, 2-amino-, ethyl ester, ethyl 2-amino-benzothiazole-6-carboxylate, ethyl 2-aminobenzo d thiazole-6-carboxylate, ethyl 2-aminobenzothiazole-6-carboxylate, ethyl2-amino-benzothiazole-6-carboxylate
ethyl 2-amino-1,3-benzothiazole-6-carboxylate

Specifications

241°C to 245°C
50850-93-6
MFCD00102724
2-amino benzo thiazole-6-carboxylic acid ethyl ester, 2-amino-6-ethoxycarbonylbenzothiazole, 2-amino-benzothiazole-6-carboxylic acid ethyl ester, 2-aminobenzothiazole-6-carboxylic acid ethyl ester, 3kid, 6-benzothiazolecarboxylic acid, 2-amino-, ethyl ester, ethyl 2-amino-benzothiazole-6-carboxylate, ethyl 2-aminobenzo d thiazole-6-carboxylate, ethyl 2-aminobenzothiazole-6-carboxylate, ethyl2-amino-benzothiazole-6-carboxylate
CCOC(=O)C1=CC2=C(C=C1)N=C(S2)N
222.262
222.26
97%
1g
C10H10N2O2S
185097
VYJSGJXWKSDUSG-UHFFFAOYSA-N
ethyl 2-amino-1,3-benzothiazole-6-carboxylate
601008
Solid
Ethyl 2-aminobenzothiazole-6-carboxylate
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures