missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 2-aminobenzothiazole-6-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H6498803

 View more versions of this product

Catalog No. AAH6498803



Specifications

Specifications

Ethyl 2-aminobenzothiazole-6-carboxylate
1g
C10H10N2O2S
185097
VYJSGJXWKSDUSG-UHFFFAOYSA-N
ethyl 2-amino-1,3-benzothiazole-6-carboxylate
601008
Solid
241°C to 245°C
50850-93-6
MFCD00102724
ethyl 2-aminobenzo d thiazole-6-carboxylate, ethyl 2-amino-benzothiazole-6-carboxylate, ethyl 2-aminobenzothiazole-6-carboxylate, 2-amino-benzothiazole-6-carboxylic acid ethyl ester, 6-benzothiazolecarboxylic acid, 2-amino-, ethyl ester, 2-amino-6-ethoxycarbonylbenzothiazole, ethyl2-amino-benzothiazole-6-carboxylate, 2-aminobenzothiazole-6-carboxylic acid ethyl ester, 2-amino benzo thiazole-6-carboxylic acid ethyl ester, 3kid
CCOC(=O)C1=CC2=C(C=C1)N=C(S2)N
222.262
222.26
97%

Chemical Identifiers

50850-93-6
222.262
VYJSGJXWKSDUSG-UHFFFAOYSA-N
601008
CCOC(=O)C1=CC2=C(C=C1)N=C(S2)N
C10H10N2O2S
MFCD00102724
ethyl 2-aminobenzo d thiazole-6-carboxylate, ethyl 2-amino-benzothiazole-6-carboxylate, ethyl 2-aminobenzothiazole-6-carboxylate, 2-amino-benzothiazole-6-carboxylic acid ethyl ester, 6-benzothiazolecarboxylic acid, 2-amino-, ethyl ester, 2-amino-6-ethoxycarbonylbenzothiazole, ethyl2-amino-benzothiazole-6-carboxylate, 2-aminobenzothiazole-6-carboxylic acid ethyl ester, 2-amino benzo thiazole-6-carboxylic acid ethyl ester, 3kid
ethyl 2-amino-1,3-benzothiazole-6-carboxylate
Documents